Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006177.1 | NP_001006177.1 | 3 - 4 | 334 | 57 | |
XM_025148593.1 | XP_025004361.1 | 3 - 4 | 387 | 57 | ||
GenBank mRNA | AJ720833.1 | - | 3 - 4 | 334 | 57 | |
CR389678.1 | - | 3 - 4 | 333 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |