Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006177.1 | NP_001006177.1 | 5 - 6 | 540 | 126 | |
XM_025148593.1 | XP_025004361.1 | 5 - 6 | 593 | 126 | ||
GenBank mRNA | AJ720833.1 | - | 5 - 6 | 540 | 126 | |
CR389678.1 | - | 5 - 6 | 539 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |