Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006202.1 | NP_001006202.1 | 9 - 10 | 1270 | 70 | |
GenBank mRNA | AJ719725.1 | - | 9 - 10 | 1270 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |