Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006211.2 | NP_001006211.1 | 4 - 5 | 667 | 88 | |
GenBank mRNA | AJ720650.1 | - | 4 - 5 | 667 | 88 | |
CR352680.1 | - | 5 - 6 | 526 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |