Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006233.1 | NP_001006233.1 | 8 - 9 | 1012 | 147 | |
GenBank mRNA | AJ720769.1 | - | 8 - 9 | 1012 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |