Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006236.1 | NP_001006236.1 | 2 - 3 | 211 | 142 | |
XM_015284675.2 | XP_015140161.1 | 2 - 3 | 244 | 142 | ||
XM_025148247.1 | XP_025004015.1 | 1 - 2 | 158 | 142 | ||
GenBank mRNA | AJ719594.1 | - | 2 - 3 | 211 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |