Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006245.1 | NP_001006245.1 | 3 - 4 | 285 | 94 | |
XM_015285752.1 | XP_015141238.1 | 4 - 5 | 617 | 94 | ||
XM_025148817.1 | XP_025004585.1 | 3 - 4 | 338 | 94 | ||
XM_025148818.1 | XP_025004586.1 | 3 - 4 | 299 | 94 | ||
XM_025148819.1 | XP_025004587.1 | 5 - 6 | 758 | 94 | ||
GenBank mRNA | AJ720178.1 | - | 3 - 4 | 285 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |