Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006245.1 | NP_001006245.1 | 6 - 7 | 706 | 86 | |
XM_015285752.1 | XP_015141238.1 | 7 - 8 | 1038 | 86 | ||
XM_025148817.1 | XP_025004585.1 | 6 - 7 | 759 | 86 | ||
XM_025148818.1 | XP_025004586.1 | 6 - 7 | 720 | 86 | ||
XM_025148819.1 | XP_025004587.1 | 8 - 9 | 1179 | 86 | ||
GenBank mRNA | AJ720178.1 | - | 6 - 7 | 706 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |