Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1H21ORF59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006258.1 | NP_001006258.1 | 6 - 7 | 829 | 120 | |
GenBank mRNA | AJ720779.1 | - | 6 - 7 | 829 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |