Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006305.2 | NP_001006305.2 | 2 - 3 | 185 | 70 | |
XM_025142771.1 | XP_024998539.1 | 2 - 3 | 289 | 70 | ||
GenBank mRNA | AJ721119.1 | - | 2 - 3 | 177 | 70 | |
CN210054.1 | - | 2 - 3 | 185 | 70 | ||
CV862347.1 | - | 1 - 2 | 118 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |