Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006305.2 | NP_001006305.2 | 5 - 6 | 498 | 101 | |
XM_025142771.1 | XP_024998539.1 | 5 - 6 | 602 | 101 | ||
GenBank mRNA | AJ721119.1 | - | 5 - 6 | 490 | 101 | |
CN210054.1 | - | 5 - 6 | 498 | 101 | ||
CV862347.1 | - | 4 - 5 | 431 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |