Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006310.2 | NP_001006310.2 | 5 - 6 | 563 | 79 | |
XM_015297547.2 | XP_015153033.2 | 5 - 6 | 1817 | 79 | ||
XM_015297549.2 | XP_015153035.2 | 5 - 6 | 797 | 79 | ||
XM_025142984.1 | XP_024998752.1 | 5 - 6 | 801 | 79 | ||
GenBank mRNA | AJ720991.1 | - | 5 - 6 | 563 | 79 | |
BU360820.1 | - | 3 - 4 | 253 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |