Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006526.1 | NP_001006526.1 | 3 - 4 | 597 | 121 | |
XM_025152207.1 | XP_025007975.1 | 4 - 5 | 645 | 121 | ||
XM_025152208.1 | XP_025007976.1 | 4 - 5 | 645 | 121 | ||
GenBank mRNA | AJ720859.1 | - | 4 - 5 | 632 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |