Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006531.1 | NP_001006531.1 | 4 - 5 | 394 | 63 | |
GenBank mRNA | AJ719915.1 | - | 4 - 5 | 394 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |