Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006545.1 | NP_001006545.1 | 6 - 7 | 1732 | 87 | |
GenBank mRNA | AJ720003.1 | - | 6 - 7 | 1732 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |