Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006561.1 | NP_001006561.1 | 4 - 5 | 401 | 71 | |
XM_015280005.2 | XP_015135491.1 | 4 - 5 | 464 | 71 | ||
XM_015280006.2 | XP_015135492.1 | 4 - 5 | 548 | 71 | ||
XM_025143404.1 | XP_024999172.1 | 4 - 5 | 540 | 71 | ||
GenBank mRNA | AJ719615.1 | - | 4 - 5 | 401 | 71 | |
BX930822.1 | - | 5 - 6 | 468 | 71 | ||
BX931149.1 | - | 4 - 5 | 428 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |