Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006574.1 | NP_001006574.1 | 9 - 10 | 1140 | 72 | |
XM_015280521.2 | XP_015136007.1 | 8 - 9 | 978 | 72 | ||
GenBank mRNA | AJ720240.1 | - | 9 - 10 | 1140 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |