Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006574.1 | NP_001006574.1 | 3 - 4 | 322 | 90 | |
XM_015280521.2 | XP_015136007.1 | 2 - 3 | 160 | 90 | ||
GenBank mRNA | AJ720240.1 | - | 3 - 4 | 322 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |