Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006574.1 | NP_001006574.1 | 5 - 6 | 495 | 120 | |
XM_015280521.2 | XP_015136007.1 | 4 - 5 | 333 | 120 | ||
GenBank mRNA | AJ720240.1 | - | 5 - 6 | 495 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |