Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007479.1 | NP_001007480.1 | 4 - 5 | 486 | 70 | |
GenBank mRNA | AJ720803.1 | - | 4 - 5 | 486 | 70 | |
AY389964.1 | - | 3 - 4 | 374 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |