Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007479.1 | NP_001007480.1 | 6 - 7 | 735 | 66 | |
GenBank mRNA | AJ720803.1 | - | 6 - 7 | 735 | 66 | |
AY389964.1 | - | 5 - 6 | 623 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |