Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TANGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007836.1 | NP_001007837.1 | 8 - 9 | 824 | 93 | |
XM_015275249.2 | XP_015130735.1 | 8 - 9 | 824 | 93 | ||
XM_015275250.2 | XP_015130736.1 | 8 - 9 | 900 | 93 | ||
XM_015275251.2 | XP_015130737.1 | 8 - 9 | 951 | 93 | ||
XM_015275252.2 | XP_015130738.1 | 8 - 9 | 1032 | 93 | ||
XM_015275253.2 | XP_015130739.1 | 8 - 9 | 964 | 93 | ||
XM_025155442.1 | XP_025011210.1 | 8 - 9 | 1956 | 93 | ||
GenBank mRNA | AJ720940.1 | - | 8 - 9 | 824 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |