Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007844.1 | NP_001007845.1 | 3 - 4 | 361 | 66 | |
XM_015295140.2 | XP_015150626.1 | 4 - 5 | 2551 | 66 | ||
XM_015295142.2 | XP_015150628.1 | 2 - 3 | 2302 | 66 | ||
GenBank mRNA | AJ719918.1 | - | 3 - 4 | 361 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |