Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007844.1 | NP_001007845.1 | 4 - 5 | 442 | 133 | |
XM_015295140.2 | XP_015150626.1 | 5 - 6 | 2632 | 133 | ||
XM_015295142.2 | XP_015150628.1 | 3 - 4 | 2383 | 133 | ||
GenBank mRNA | AJ719918.1 | - | 4 - 5 | 442 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |