Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR1OP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007855.1 | NP_001007856.1 | 2 - 3 | 198 | 110 | |
GenBank mRNA | AJ720090.1 | - | 2 - 3 | 198 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |