Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007905.2 | NP_001007906.1 | 2 - 3 | 497 | 62 | |
GenBank mRNA | BX933632.1 | - | 2 - 3 | 233 | 62 | |
BX933635.1 | - | 2 - 3 | 250 | 62 | ||
BX933691.1 | - | 2 - 3 | 231 | 62 | ||
BX934589.1 | - | 2 - 3 | 252 | 62 | ||
BX935045.2 | - | 2 - 3 | 239 | 62 | ||
BX935881.1 | - | 2 - 3 | 237 | 62 | ||
BX935913.1 | - | 2 - 3 | 493 | 62 | ||
BX950755.2 | - | 2 - 3 | 231 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |