Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008453.1 | NP_001008453.1 | 305 | 79 | ||
GenBank mRNA | AJ720095.1 | - | 305 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus |