Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204757.1 | NP_990088.1 | 2 - 3 | 141 | 61 | |
XM_015284449.2 | XP_015139935.1 | 2 - 3 | 163 | 61 | ||
GenBank mRNA | D16184.1 | - | 2 - 3 | 141 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |