Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204768.1 | NP_990099.1 | 4 - 5 | 819 | 71 | |
GenBank mRNA | AF178671.1 | - | 4 - 5 | 819 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |