Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204783.1 | NP_990114.1 | 3 - 4 | 633 | 97 | |
GenBank mRNA | D31900.1 | - | 3 - 4 | 633 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |