Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205433.1 | NP_990764.1 | 2 - 3 | 387 | 105 | |
GenBank mRNA | M95706.1 | - | 2 - 3 | 258 | 105 | |
M95707.1 | - | 2 - 3 | 387 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |