Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205451.1 | NP_990782.1 | 3 - 4 | 443 | 68 | |
GenBank mRNA | M11314.1 | - | 3 - 4 | 443 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |