Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204445.2 | NP_989776.1 | 2 - 3 | 300 | 156 | |
XM_025141544.1 | XP_024997312.1 | 2 - 3 | 337 | 156 | ||
GenBank mRNA | AY226461.1 | - | 2 - 3 | 302 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |