Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101831.1 | NP_001095301.1 | 2 | 152 | 151 | |
GenBank mRNA | AF123456.2 | - | 2 | 244 | 151 | |
AF211349.1 | - | 2 | 244 | 151 | ||
AY448019.1 | - | 2 | 152 | 151 | ||
AY448023.1 | - | 2 | 152 | 151 | ||
AY448024.1 | - | 2 | 152 | 151 | ||
DQ164800.1 | - | 2 | 152 | 151 | ||
GU396269.1 | - | 2 | 437 | 151 | ||
HQ127225.1 | - | 2 | 354 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |