Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRFPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127170.1 | NP_001120642.1 | 1 - 2 | 344 | 84 | |
XM_015276274.2 | XP_015131760.1 | 1 - 2 | 470 | 84 | ||
GenBank mRNA | EU525171.1 | - | 1 - 2 | 344 | 84 | |
KY856722.1 | - | 1 - 2 | 482 | 84 | ||
KY856723.1 | - | 1 - 2 | 482 | 84 | ||
KY856724.1 | - | 1 - 2 | 482 | 84 | ||
KY856725.1 | - | 1 - 2 | 482 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Gallus gallus Probe spans exons |