Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130743.1 | NP_001124215.1 | 4 - 5 | 512 | 80 | |
XM_015285151.2 | XP_015140637.1 | 3 - 4 | 882 | 80 | ||
GenBank mRNA | AB368182.1 | - | 4 - 5 | 512 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Gallus gallus Probe spans exons |