Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135550.1 | NP_001129022.1 | 3 - 4 | 328 | 147 | |
XM_015290135.2 | XP_015145621.1 | 3 - 4 | 339 | 147 | ||
XM_015290146.2 | XP_015145632.1 | 3 - 4 | 332 | 147 | ||
XM_025148902.1 | XP_025004670.1 | 3 - 4 | 325 | 147 | ||
XM_025148903.1 | XP_025004671.1 | 3 - 4 | 281 | 147 | ||
XM_025148905.1 | XP_025004673.1 | 3 - 4 | 521 | 147 | ||
XM_025148906.1 | XP_025004674.1 | 3 - 4 | 360 | 147 | ||
XM_025148908.1 | XP_025004676.1 | 3 - 4 | 512 | 147 | ||
GenBank mRNA | EF585397.1 | - | 3 - 4 | 314 | 147 | |
EF585398.1 | - | 3 - 4 | 328 | 147 | ||
EU309478.1 | - | 3 - 4 | 316 | 147 | ||
EU309479.1 | - | 3 - 4 | 326 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |