Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145200.1 | NP_001138672.1 | 4 - 5 | 237 | 74 | |
XM_025151721.1 | XP_025007489.1 | 4 - 5 | 256 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |