Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145226.2 | NP_001138698.2 | 2 - 3 | 203 | 60 | |
XM_015296218.2 | XP_015151704.1 | 3 - 4 | 276 | 60 | ||
XM_015296220.2 | XP_015151706.1 | 3 - 4 | 753 | 60 | ||
XM_015296221.2 | XP_015151707.1 | 3 - 4 | 268 | 60 | ||
XM_015296222.2 | XP_015151708.1 | 3 - 4 | 278 | 60 | ||
GenBank mRNA | FM200428.1 | - | 2 - 3 | 203 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |