Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRIMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145492.1 | NP_001138964.1 | 1 - 2 | 95 | 86 | |
XM_015287567.2 | XP_015143053.1 | 1 - 2 | 300 | 86 | ||
GenBank mRNA | FJ611893.1 | - | 1 - 2 | 95 | 86 | |
FJ611894.1 | - | 1 - 2 | 95 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |