Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146132.1 | NP_001139604.1 | 2 - 3 | 788 | 103 | |
GenBank mRNA | AM933592.1 | - | 2 - 3 | 788 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |