Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204844.2 | NP_990175.2 | 1 | 2277 | 73 | |
GenBank mRNA | AF151734.1 | - | 1 | 2142 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |