Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF039851.1 | - | 15 - 16 | 1962 | 76 | |
AK287031.1 | - | 24 - 25 | 3786 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |