Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF039851.1 | - | 21 - 22 | 2751 | 62 | |
AK287031.1 | - | 30 - 31 | 4575 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Glycine max Probe spans exons |