Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251128.1 | NP_001238057.1 | 5 - 6 | 922 | 82 | |
GenBank mRNA | AF126550.1 | - | 5 - 6 | 922 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |