Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250643.2 | NP_001237572.1 | 2 - 3 | 652 | 90 | |
GenBank mRNA | AF141602.1 | - | 2 - 3 | 657 | 90 | |
AK244280.1 | - | 2 - 3 | 675 | 90 | ||
AK286711.1 | - | 2 - 3 | 682 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |