Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250643.2 | NP_001237572.1 | 3 - 4 | 729 | 75 | |
GenBank mRNA | AF141602.1 | - | 3 - 4 | 734 | 75 | |
AK244280.1 | - | 3 - 4 | 752 | 75 | ||
AK286711.1 | - | 3 - 4 | 759 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |