Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248230.1 | NP_001235159.1 | 6 - 7 | 467 | 95 | |
GenBank mRNA | AF255338.1 | - | 6 - 7 | 467 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Protease Recommended Assay Glycine max Probe spans exons |