Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC548033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250806.1 | NP_001237735.1 | 1 - 2 | 308 | 62 | |
GenBank mRNA | AF260565.1 | - | 1 - 2 | 308 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Glycine max Probe spans exons |