Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248749.2 | NP_001235678.1 | 3 - 4 | 263 | 71 | |
GenBank mRNA | AF464906.1 | - | 3 - 4 | 301 | 71 | |
AK244706.1 | - | 3 - 4 | 304 | 71 | ||
BT089816.1 | - | 3 - 4 | 265 | 71 | ||
BT092715.1 | - | 3 - 4 | 307 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |